Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032259.3 | NP_115635.1 | 2 | 1334 | ||
XM_011522699.2 | XP_011521001.1 | 3 | 1838 | |||
GenBank mRNA | AK301146.1 | - | 2 | 1318 | ||
AL136863.1 | - | 2 | 1311 | |||
BC008025.1 | - | 2 | 799 | 3544791 | ||
BC009761.2 | - | 2 | 334 |