Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005054.2 | NP_005045.2 | 17 | 2497 | ||
NM_032260.2 | NP_115636.1 | 18 | 2953 | |||
XM_006712794.3 | XP_006712857.2 | 17 | 2528 | |||
XM_017005080.1 | XP_016860569.1 | 17 | 2517 | |||
XM_017005081.1 | XP_016860570.1 | 17 | 2528 | |||
XM_017005082.1 | XP_016860571.1 | 15 | 2199 | |||
GenBank mRNA | AL136868.1 | - | 18 | 2954 | ||
EU794682.1 | - | 17 | 2497 | |||
U64675.2 | - | 17 | 2497 |
Gene Symbol: RGPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037866.1 | NP_001032955.1 | 18 | 2953 | ||
NM_001123363.3 | NP_001116835.1 | 18 | 2953 | |||
XM_011511766.2 | XP_011510068.1 | 17 | 2537 | |||
XM_011511767.2 | XP_011510069.1 | 17 | 2537 | |||
XM_011511769.2 | XP_011510071.1 | 17 | 2537 | |||
XM_011511770.2 | XP_011510072.1 | 17 | 2537 | |||
XM_017004839.1 | XP_016860328.1 | 17 | 2537 | |||
XM_017004840.1 | XP_016860329.1 | 17 | 2610 | |||
XM_017004841.1 | XP_016860330.1 | 17 | 2740 | |||
XM_017004842.1 | XP_016860331.1 | 17 | 2517 | |||
XM_017004843.1 | XP_016860332.1 | 17 | 2537 | |||
XM_017004844.1 | XP_016860333.1 | 15 | 2199 | |||
GenBank mRNA | AK056675.1 | - | 13 | 1811 |
Gene Symbol: RGPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164463.1 | NP_001157935.1 | 17 | 2612 | ||
XM_005263747.4 | XP_005263804.1 | 17 | 2610 | |||
XM_011511733.2 | XP_011510035.1 | 17 | 2610 | |||
XM_017004825.1 | XP_016860314.1 | 17 | 2610 | |||
XM_017004826.1 | XP_016860315.1 | 17 | 2610 |