Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134435.2 | NP_001127907.1 | 3 | 232 | ||
NM_001323027.1 | NP_001309956.1 | 3 | 621 | |||
NM_001323028.1 | NP_001309957.1 | 3 | 444 | |||
NM_001323029.1 | NP_001309958.1 | 4 | 511 | |||
NM_001323030.1 | NP_001309959.1 | 4 | 309 | |||
NM_032263.4 | NP_115639.1 | 4 | 511 | |||
XM_017007316.1 | XP_016862805.1 | 4 | 501 | |||
XM_017007317.1 | XP_016862806.1 | 3 | 232 | |||
GenBank mRNA | AL136889.1 | - | 4 | 495 | ||
BC016156.1 | - | 4 | 459 | |||
BC136253.1 | - | 3 | 177 | 9020240 | ||
CK001737.1 | - | 4 | 514 | |||
CR533515.1 | - | 1 | 86 | |||
DA358032.1 | - | 3 | 229 | |||
DA566107.1 | - | 3 | 232 |