Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256416.2 | NP_001243345.1 | 7 | 1246 | ||
NM_001256417.2 | NP_001243346.1 | 6 | 1036 | |||
NM_032264.4 | NP_115640.1 | 7 | 1246 | |||
NR_046176.2 | 10 | 1775 | ||||
XM_006710957.1 | XP_006711020.1 | 7 | 1215 | |||
XM_006710958.1 | XP_006711021.1 | 10 | 1560 | |||
XM_006710959.2 | XP_006711022.1 | 8 | 1344 | |||
XM_011542276.1 | XP_011540578.1 | 11 | 1817 | |||
XM_011542277.1 | XP_011540579.1 | 11 | 1802 | |||
XM_011542278.1 | XP_011540580.1 | 11 | 1631 | |||
XM_011542281.1 | XP_011540583.1 | 8 | 1167 | |||
XM_017002498.1 | XP_016857987.1 | 11 | 1616 | |||
XM_017002499.1 | XP_016857988.1 | 6 | 882 | |||
XM_017002500.1 | XP_016857989.1 | 10 | 1746 | |||
XM_017002501.1 | XP_016857990.1 | 11 | 1631 | |||
XM_017002502.1 | XP_016857991.1 | 10 | 1560 | |||
GenBank mRNA | AK095602.1 | - | 10 | 1728 | ||
AK292580.1 | - | 7 | 1111 | |||
AK299840.1 | - | 6 | 998 | |||
AK314597.1 | - | 7 | 1174 | |||
AL136890.1 | - | 7 | 1023 | |||
AL834330.1 | - | 3 | 219 | |||
AY598326.1 | - | 6 | 848 | |||
AY894564.1 | - | 6 | 752 | |||
BC024011.2 | - | 7 | 1173 | 4827023 |