Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136046.2 | NP_001129518.1 | 3 | 691 | ||
NM_001267822.1 | NP_001254751.1 | 3 | 691 | |||
NM_032265.2 | NP_115641.1 | 3 | 691 | |||
GenBank mRNA | AK302181.1 | - | 3 | 682 | ||
AK310469.1 | - | 2 | 1144 | |||
AL136893.1 | - | 2 | 664 |