Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032271.2 | NP_115647.2 | 11, 12 | 1177 | ||
XM_005255627.4 | XP_005255684.1 | 11, 12 | 1217 | |||
XM_011522700.1 | XP_011521002.1 | 11, 12 | 1219 | |||
GenBank mRNA | AK090624.1 | - | 11, 12 | 1191 | ||
AL136921.1 | - | 10, 11 | 1073 | |||
AY569455.1 | - | 10, 11 | 1130 | |||
BC024267.1 | - | 9, 10 | 963 |