Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032289.2 | NP_115665.1 | 12 | 1982 | ||
XM_011537696.2 | XP_011535998.1 | 13 | 2038 | |||
XM_017009975.1 | XP_016865464.1 | 13 | 3345 | |||
XM_017009976.1 | XP_016865465.1 | 13 | 4153 | |||
XM_017009977.1 | XP_016865466.1 | 13 | 4033 | |||
GenBank mRNA | AL136559.1 | - | 12 | 1982 | ||
AL834268.1 | - | 8 | 764 | |||
BC038233.1 | - | 12 | 1959 | 5752229 |