Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GARNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286779.1 | NP_001273708.1 | 16, 17 | 1618 | ||
NM_032293.4 | NP_115669.3 | 15, 16 | 1755 | |||
NR_104591.1 | 15, 16 | 1755 | ||||
XM_005252267.3 | XP_005252324.1 | 17, 18 | 1610 | |||
XM_005252268.2 | XP_005252325.1 | 17, 18 | 1672 | |||
XM_011519086.2 | XP_011517388.1 | 18, 19 | 1682 | |||
XM_011519087.2 | XP_011517389.1 | 18, 19 | 1665 | |||
XM_011519089.2 | XP_011517391.1 | 17, 18 | 1568 | |||
XM_011519090.2 | XP_011517392.1 | 16, 17 | 1451 | |||
XM_017015202.1 | XP_016870691.1 | 16, 17 | 1648 | |||
GenBank mRNA | AK293732.1 | - | 15, 16 | 1653 | ||
AK294970.1 | - | 15, 16 | 1257 | |||
AK296246.1 | - | 16, 17 | 1618 | |||
AL136573.1 | - | 15, 16 | 1309 | |||
BC034983.2 | - | 15, 16 | 1365 | 4821143 |