Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143852.1 | NP_001137324.1 | 4 | 589 | ||
NM_032300.4 | NP_115676.1 | 4 | 650 | |||
XM_011538836.2 | XP_011537138.1 | 4 | 925 | |||
XM_011538837.2 | XP_011537139.1 | 4 | 498 | |||
XM_017020024.1 | XP_016875513.1 | 4 | 589 | |||
XM_017020025.1 | XP_016875514.1 | 4 | 613 | |||
GenBank mRNA | AK092736.1 | - | 4 | 589 | ||
AK301262.1 | - | 4 | 567 | |||
AY007230.1 | - | 4 | 651 | |||
BC004285.1 | - | 4 | 570 | 3617849 | ||
BC022838.1 | - | 4 | 879 |