Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195822.1 | NP_001182751.1 | 2 | 331 | ||
NM_032303.4 | NP_115679.2 | 2 | 331 | |||
XM_011519091.2 | XP_011517393.1 | 2 | 331 | |||
XM_017015204.1 | XP_016870693.1 | 2 | 948 | |||
GenBank mRNA | AK098721.1 | - | 2 | 302 | ||
AK225946.1 | - | 2 | 132 | |||
AK289929.1 | - | 2 | 159 | |||
AK292486.1 | - | 2 | 138 | |||
AK301469.1 | - | 2 | 128 | |||
AK313605.1 | - | 2 | 138 | |||
AY093428.1 | - | 2 | 170 | |||
BC004331.1 | - | 2 | 160 | 3630835 | ||
BC095451.1 | - | 2 | 108 | 30405073 | ||
DB052480.1 | - | 2 | 335 |