Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290137.1 | NP_001277066.1 | 9 | 1323 | ||
NM_001290139.1 | NP_001277068.1 | 9 | 1393 | |||
NM_001323635.1 | NP_001310564.1 | 9 | 1428 | |||
NM_001323636.1 | NP_001310565.1 | 8 | 1348 | |||
NM_032304.3 | NP_115680.1 | 8 | 1354 | |||
XM_005255632.1 | XP_005255689.1 | 8 | 1561 | |||
XM_011522712.1 | XP_011521014.1 | 8 | 1585 | |||
XM_017023774.1 | XP_016879263.1 | 8 | 1417 | |||
GenBank mRNA | AK054841.1 | - | 6 | 1649 | ||
AK098340.1 | - | 3 | 2181 | |||
BC015008.1 | - | 8 | 1282 | 4862387 | ||
BC033796.1 | - | 7 | 1683 | 5173915 | ||
BC036747.1 | - | 7 | 1683 | |||
BQ067068.1 | - | 4 | 641 |