Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318010.1 | NP_001304939.1 | 2 | 350 | ||
NM_001318011.1 | NP_001304940.1 | 2 | 350 | |||
NM_032310.4 | NP_115686.3 | 2 | 350 | |||
NR_134464.1 | 2 | 350 | ||||
NR_134465.1 | 2 | 350 | ||||
NR_134466.1 | 2 | 350 | ||||
NR_134467.1 | 2 | 350 | ||||
NR_134468.1 | 2 | 350 | ||||
GenBank mRNA | AK057716.1 | - | 2 | 256 | ||
AK091611.1 | - | 2 | 256 | |||
BC004500.2 | - | 3 | 219 | 3833318 | ||
BC038856.1 | - | 2 | 243 | 5215640 | ||
BC091476.1 | - | 2 | 193 | |||
BM930683.1 | - | 2 | 195 | |||
BX354102.2 | - | 2 | 228 | |||
CD676329.1 | - | 2 | 206 | |||
DN998463.1 | - | 2 | 243 | |||
HY005780.1 | - | 2 | 351 |