Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297741.1 | NP_001284670.1 | 8 | 1346 | ||
NM_001297742.1 | NP_001284671.1 | 8 | 1536 | |||
NM_032320.6 | NP_115696.2 | 9 | 1504 | |||
XM_017018405.1 | XP_016873894.1 | 9 | 2620 | |||
XM_017018406.1 | XP_016873895.1 | 10 | 1599 | |||
XM_017018407.1 | XP_016873896.1 | 10 | 1579 | |||
XM_017018408.1 | XP_016873897.1 | 9 | 1474 | |||
XM_017018409.1 | XP_016873898.1 | 9 | 1446 | |||
XM_017018410.1 | XP_016873899.1 | 9 | 1438 | |||
XM_017018411.1 | XP_016873900.1 | 8 | 1333 | |||
GenBank mRNA | AF329102.2 | - | 9 | 1504 | ||
AK095946.1 | - | 8 | 1344 | |||
AK294259.1 | - | 8 | 1325 | |||
AK298226.1 | - | 8 | 1221 | |||
AK316163.1 | - | 8 | 1536 | |||
AL832981.1 | - | 9 | 1485 | |||
AY221959.2 | - | 9 | 1547 | |||
BC005071.1 | - | 9 | 1466 | 3531347 | ||
BC017072.1 | - | 4 | 481 | |||
BC062633.1 | - | 5 | 614 |