Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042519.1 | NP_001035984.1 | 2 | 580 | ||
NM_001042520.1 | NP_001035985.1 | 2 | 536 | |||
NM_001042521.1 | NP_001035986.1 | 3 | 548 | |||
NM_032321.2 | NP_115697.2 | 2 | 507 | |||
XM_005246905.1 | XP_005246962.1 | 4 | 729 | |||
XM_006712795.2 | XP_006712858.1 | 2 | 414 | |||
XM_011511982.1 | XP_011510284.1 | 5 | 816 | |||
XM_011511983.1 | XP_011510285.1 | 5 | 881 | |||
XM_011511984.2 | XP_011510286.1 | 5 | 672 | |||
XM_011511985.2 | XP_011510287.1 | 4 | 588 | |||
XM_011511986.2 | XP_011510288.1 | 3 | 752 | |||
XM_017005096.1 | XP_016860585.1 | 2 | 596 | |||
GenBank mRNA | AK075168.1 | - | 2 | 1262 | ||
AK313395.1 | - | 2 | 476 | |||
BX648070.1 | - | 1 | 85 | |||
DA389238.1 | - | 2 | 537 |