Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184992.1 | NP_001171921.1 | 2 | 570 | ||
NM_032322.3 | NP_115698.3 | 2 | 570 | |||
NM_197939.1 | NP_922921.1 | 2 | 570 | |||
XM_005258043.4 | XP_005258100.1 | 3 | 2082 | |||
XM_017025223.1 | XP_016880712.1 | 2 | 756 | |||
GenBank mRNA | AB470605.1 | - | 2 | 434 | ||
AJ496729.1 | - | 2 | 485 | |||
AK122646.1 | - | 2 | 443 | |||
AK122709.1 | - | 3 | 365 | |||
AK312979.1 | - | 2 | 446 | |||
AY598332.1 | - | 2 | 434 | |||
BC005084.1 | - | 2 | 485 | |||
BC082262.1 | - | 2 | 448 | |||
BC126420.1 | - | 2 | 524 | 8992136 | ||
BC126422.1 | - | 2 | 524 | 8992138 |