Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015877.1 | NP_001015877.1 | 4 | 537 | ||
NM_032335.3 | NP_115711.2 | 4 | 537 | |||
NM_032458.2 | NP_115834.1 | 4 | 537 | |||
GenBank mRNA | AB058726.1 | - | 4 | 423 | ||
AK290095.1 | - | 4 | 508 | |||
AK303369.1 | - | 3 | 400 | |||
AL832725.1 | - | 4 | 530 | |||
AY157622.1 | - | 4 | 537 | |||
BC005994.1 | - | 4 | 476 | 4064169 | ||
CD685336.1 | - | 4 | 545 | |||
DR000282.1 | - | 4 | 572 |