Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303107.1 | NP_001290036.1 | 3 | 953 | ||
NM_001303108.1 | NP_001290037.1 | 2 | 828 | |||
NM_032342.2 | NP_115718.1 | 2 | 898 | |||
XM_017015206.1 | XP_016870695.1 | 2 | 1088 | |||
GenBank mRNA | AL521069.3 | - | 3 | 855 | ||
BC006115.1 | - | 2 | 870 | 3503592 | ||
BC033550.1 | - | 4 | 1696 | 4822472 | ||
BC113967.1 | - | 1 | 75 |