Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320610.1 | NP_001307539.1 | 1, 2 | 180 | ||
NM_032343.2 | NP_115719.1 | 1, 2 | 180 | |||
XM_011513235.2 | XP_011511537.1 | 1, 2 | 159 | |||
XM_017007320.1 | XP_016862809.1 | 1, 2 | 159 | |||
XM_017007321.1 | XP_016862810.1 | 1, 2 | 159 | |||
XM_017007322.1 | XP_016862811.1 | 1, 2 | 159 | |||
GenBank mRNA | AK310910.1 | - | 1, 2 | 159 | ||
AK311056.1 | - | 1, 2 | 169 | |||
BC006123.1 | - | 1, 2 | 125 | 3535457 | ||
DN992600.1 | - | 1, 2 | 187 | |||
JF264889.1 | - | 1, 2 | 125 |