Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032346.1 | NP_115722.1 | 6 | 930 | ||
GenBank mRNA | BC006146.1 | - | 6 | 930 | 3944994 |