Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032351.5 | NP_115727.5 | 6 | 717 | ||
GenBank mRNA | AK172741.1 | - | 6 | 577 | ||
AK225031.1 | - | 6 | 577 | |||
AK293605.1 | - | 6 | 683 | |||
BC130382.1 | - | 6 | 608 | 40146413 | ||
BC130384.1 | - | 6 | 608 | 40146415 | ||
BG761720.1 | - | 6 | 577 |