Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032354.3 | NP_115730.2 | 5 | 1348 | ||
NM_183065.2 | NP_898888.1 | 5 | 1330 | |||
GenBank mRNA | BC006292.2 | - | 2 | 1021 |