Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130146.1 | NP_001123618.1 | 8 | 839 | ||
NM_001130147.1 | NP_001123619.1 | 8 | 866 | |||
NM_001130148.1 | NP_001123620.1 | 7 | 812 | |||
NM_032358.3 | NP_115734.1 | 9 | 893 | |||
GenBank mRNA | AK027638.1 | - | 9 | 811 | ||
AK293165.1 | - | 8 | 777 | |||
AK308216.1 | - | 8 | 821 | |||
AK316029.1 | - | 7 | 814 | |||
BC006444.2 | - | 8 | 782 | 3616619 | ||
CX784327.1 | - | 8 | 746 |