Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167924.1 | NP_001161396.1 | 9 | 734 | ||
NM_032359.3 | NP_115735.2 | 9 | 875 | |||
GenBank mRNA | AK124387.1 | - | 9 | 843 | ||
AK291392.1 | - | 9 | 847 | |||
AK300704.1 | - | 9 | 736 | |||
AK312483.1 | - | 9 | 875 | |||
BC006475.1 | - | 9 | 872 | 2819663 | ||
BC006512.2 | - | 9 | 872 | 2819663 |