Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032361.3 | NP_115737.1 | 5 | 965 | ||
XM_017009985.1 | XP_016865474.1 | 6 | 832 | |||
GenBank mRNA | BC006849.1 | - | 5 | 927 | 3451612 | |
BC066325.1 | - | 5 | 922 | 5303134 | ||
BC068499.1 | - | 5 | 884 | 5262880 | ||
BU781626.1 | - | 4 | 562 |
Gene Symbol: LOC728554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003615.2 | 5 | 1191 | |||
GenBank mRNA | AK310274.1 | - | 5 | 665 |