Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032361.3 | NP_115737.1 | 6 | 1093 | ||
XM_017009985.1 | XP_016865474.1 | 7 | 960 | |||
GenBank mRNA | BC006849.1 | - | 6 | 1055 | 3451612 | |
BC068499.1 | - | 6 | 1012 | 5262880 |
Gene Symbol: LOC728554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003615.2 | 6 | 1319 |