Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033082.3 | NP_149073.1 | 2 | 156 | ||
NR_026722.1 | 2 | 156 | ||||
NR_026723.1 | 2 | 156 | ||||
GenBank mRNA | AF161434.1 | - | 2 | 123 | ||
AF486281.1 | - | 2 | 101 | |||
AK222719.1 | - | 2 | 134 | |||
AK290508.1 | - | 2 | 124 | |||
BC007099.1 | - | 2 | 122 | 4273903 | ||
BC093051.1 | - | 2 | 122 | 30382893 | ||
BI520421.1 | - | 2 | 156 |