Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304813.1 | NP_001291742.1 | 3, 4 | 467 | ||
NM_001304814.1 | NP_001291743.1 | 3, 4 | 564 | |||
NM_032376.3 | NP_115752.1 | 2, 3 | 396 | |||
XM_005257737.2 | XP_005257794.1 | 3, 4 | 1744 | |||
XM_011525346.1 | XP_011523648.1 | 5, 6 | 1981 | |||
XM_011525347.1 | XP_011523649.1 | 4, 5 | 1858 | |||
XM_011525349.1 | XP_011523651.1 | 3, 4 | 1764 | |||
XM_011525351.1 | XP_011523653.1 | 3, 4 | 1211 | |||
XM_011525353.1 | XP_011523655.1 | 3, 4 | 1623 | |||
GenBank mRNA | AB097008.1 | - | 2, 3 | 332 | ||
AK130810.1 | - | 2, 3 | 330 | |||
AK172826.1 | - | 2, 3 | 348 | |||
AK294237.1 | - | 2, 3 | 247 | |||
AK313855.1 | - | 2, 3 | 330 | |||
BC007438.2 | - | 2, 3 | 344 | 3028940 | ||
CN261562.1 | - | 2, 3 | 397 | |||
DC311483.1 | - | 2, 3 | 395 | |||
DC393428.1 | - | 3, 4 | 455 |