Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304813.1 | NP_001291742.1 | 3 | 313 | ||
NM_001304814.1 | NP_001291743.1 | 3 | 410 | |||
NM_032376.3 | NP_115752.1 | 2 | 242 | |||
XM_005257737.2 | XP_005257794.1 | 3 | 1590 | |||
XM_011525346.1 | XP_011523648.1 | 5 | 1827 | |||
XM_011525347.1 | XP_011523649.1 | 4 | 1704 | |||
XM_011525349.1 | XP_011523651.1 | 3 | 1610 | |||
XM_011525351.1 | XP_011523653.1 | 3 | 1057 | |||
XM_011525353.1 | XP_011523655.1 | 3 | 1469 | |||
GenBank mRNA | AB097008.1 | - | 2 | 178 | ||
AK130810.1 | - | 2 | 176 | |||
AK172826.1 | - | 2 | 194 | |||
AK294237.1 | - | 2 | 93 | |||
AK313855.1 | - | 2 | 176 | |||
BC007438.2 | - | 2 | 190 | 3028940 | ||
CN261562.1 | - | 2 | 243 | |||
DC311483.1 | - | 2 | 241 | |||
DC344561.1 | - | 3 | 412 | |||
DC393428.1 | - | 3 | 301 | |||
DC393490.1 | - | 3 | 481 |