Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032377.3 | NP_115753.1 | 4 | 986 | ||
XM_017027356.1 | XP_016882845.1 | 5 | 1141 | |||
XM_017027357.1 | XP_016882846.1 | 4 | 1000 | |||
XM_017027358.1 | XP_016882847.1 | 4 | 1140 | |||
XM_017027359.1 | XP_016882848.1 | 4 | 1056 | |||
GenBank mRNA | AK001171.1 | - | 5 | 1137 | ||
BC007516.2 | - | 3 | 935 | 3028469 | ||
BG424379.1 | - | 467 |