Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281302.1 | NP_001268231.1 | 4, 5 | 726 | ||
NM_032380.4 | NP_115756.2 | 3, 4 | 608 | |||
NM_170681.2 | NP_733781.1 | 3, 4 | 608 | |||
NM_170691.2 | NP_733792.1 | 3, 4 | 608 | |||
NR_104006.1 | 3, 4 | 608 | ||||
XM_011543691.2 | XP_011541993.1 | 3, 4 | 391 | |||
XM_017009986.1 | XP_016865475.1 | 3, 4 | 502 | |||
GenBank mRNA | AF111808.1 | - | 3, 4 | 358 | ||
AF367997.1 | - | 3, 4 | 454 | |||
AK025314.1 | - | 3, 4 | 385 | |||
AK307839.1 | - | 3, 4 | 298 | |||
AK308422.1 | - | 3, 4 | 355 | |||
AK315705.1 | - | 3, 4 | 387 | |||
AY453587.1 | - | 3, 4 | 352 | |||
BC015712.1 | - | 4, 5 | 460 | 3922036 | ||
BC030612.1 | - | 3, 4 | 357 | 4811939 | ||
BI828937.1 | - | 3, 4 | 611 |