Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318099.1 | NP_001305028.1 | 5 | 1937 | ||
NM_001318100.1 | NP_001305029.1 | 5 | 2014 | |||
NM_001318101.1 | NP_001305030.1 | 3 | 1215 | |||
NM_032429.3 | NP_115805.1 | 5 | 2044 | |||
XM_005270222.4 | XP_005270279.1 | 5 | 2276 | |||
XM_017016780.1 | XP_016872269.1 | 4 | 4405 | |||
XM_017016781.1 | XP_016872270.1 | 4 | 1385 | |||
GenBank mRNA | AB058716.1 | - | 4 | 4896 | ||
AL834338.1 | - | 3 | 1420 | |||
AY029201.1 | - | 4 | 1758 | |||
BC005855.2 | - | 2 | 748 | |||
BC006212.2 | - | 5 | 1902 | 3161855 | ||
BC058938.1 | - | 5 | 1977 | 3913470 |