Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001344.3 | NP_001335.1 | 2 | 358 | ||
GenBank mRNA | BC007403.2 | - | 2 | 330 | 2988797 | |
BC009798.1 | - | 2 | 332 | 4284779 | ||
BP198316.1 | - | 2 | 358 | |||
CR407682.1 | - | 2 | 278 | |||
CR542204.1 | - | 2 | 278 | |||
D15057.1 | - | 2 | 344 |