Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYVN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032431.2 | NP_115807.1 | 5, 6 | 521 | ||
NM_172230.2 | NP_757385.1 | 5, 6 | 521 | |||
XM_011545303.2 | XP_011543605.1 | 5, 6 | 1369 | |||
GenBank mRNA | AK126912.1 | - | 5, 6 | 522 | ||
AL834262.1 | - | 5, 6 | 521 | |||
BC030530.1 | - | 5, 6 | 462 | 5211742 |