Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM2 SIRNA Assays ›
Gene Symbol:
ABLIM2
Gene Name:
actin binding LIM protein family member 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 7965310 - 8158832 on Build GRCh38
UniGene ID:
Hs.233404
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s39022
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130083.1
NP_001123555.1
10
1092
NM_001130084.1
NP_001123556.1
10
1092
NM_001130085.1
NP_001123557.1
10
1092
NM_001130086.1
NP_001123558.1
10
1092
NM_001130087.1
NP_001123559.1
10
1092
NM_001130088.1
NP_001123560.1
10
1092
NM_001286688.1
NP_001273617.1
5
522
NM_032432.4
NP_115808.3
10
1092
XM_005248014.4
XP_005248071.1
10
1063
XM_005248017.4
XP_005248074.1
10
1063
XM_005248018.4
XP_005248075.1
10
1062
XM_005248019.4
XP_005248076.1
10
1062
XM_005248020.4
XP_005248077.1
10
1062
XM_005248021.4
XP_005248078.1
10
1061
XM_005248022.4
XP_005248079.1
10
1062
XM_005248023.4
XP_005248080.1
10
1062
XM_005248024.4
XP_005248081.1
10
1061
XM_005248027.4
XP_005248084.1
10
1061
XM_005248028.4
XP_005248085.1
10
1058
XM_005248029.4
XP_005248086.1
10
1060
XM_005248030.4
XP_005248087.1
10
1060
XM_005248031.4
XP_005248088.1
10
1062
XM_006713923.3
XP_006713986.1
10
1063
XM_006713924.3
XP_006713987.1
10
1062
XM_006713925.3
XP_006713988.1
10
1063
XM_006713927.3
XP_006713990.1
10
1061
XM_011513586.2
XP_011511888.1
10
1067
XM_017008706.1
XP_016864195.1
10
1064
XM_017008707.1
XP_016864196.1
10
1073
XM_017008708.1
XP_016864197.1
10
1062
XM_017008709.1
XP_016864198.1
10
1063
XM_017008710.1
XP_016864199.1
10
1062
XM_017008711.1
XP_016864200.1
10
1062
XM_017008712.1
XP_016864201.1
10
1062
XM_017008713.1
XP_016864202.1
10
1061
XM_017008714.1
XP_016864203.1
10
1073
XM_017008715.1
XP_016864204.1
10
1062
XM_017008716.1
XP_016864205.1
10
1061
XM_017008717.1
XP_016864206.1
10
1061
XM_017008718.1
XP_016864207.1
10
1073
XM_017008719.1
XP_016864208.1
10
1061
XM_017008720.1
XP_016864209.1
10
1060
XM_017008721.1
XP_016864210.1
10
1060
XM_017008722.1
XP_016864211.1
10
1061
XM_017008723.1
XP_016864212.1
10
1073
XM_017008724.1
XP_016864213.1
10
1073
XM_017008725.1
XP_016864214.1
10
1073
XM_017008726.1
XP_016864215.1
5
522
XM_017008727.1
XP_016864216.1
5
522
GenBank mRNA
AB058711.1
-
9
851
AJ748600.1
-
10
1014
AJ748601.1
-
10
1014
AK094754.1
-
10
1071
AK094798.1
-
5
492
AK301060.1
-
10
1055
BC067214.1
-
10
1104
BC122567.1
-
10
1043
40085570
DA392726.1
-
5
522
DQ413176.1
-
10
969
DQ413177.1
-
9
949
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
actin cytoskeleton organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0