Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM2 SIRNA Assays ›
Gene Symbol:
ABLIM2
Gene Name:
actin binding LIM protein family member 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 7965310 - 8158832 on Build GRCh38
UniGene ID:
Hs.233404
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s39024
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130083.1
NP_001123555.1
10
1135
NM_001130084.1
NP_001123556.1
10
1135
NM_001130085.1
NP_001123557.1
10
1135
NM_001130086.1
NP_001123558.1
10
1135
NM_001130087.1
NP_001123559.1
10
1135
NM_001130088.1
NP_001123560.1
10
1135
NM_001286688.1
NP_001273617.1
5
565
NM_032432.4
NP_115808.3
10
1135
XM_005248014.4
XP_005248071.1
10
1106
XM_005248017.4
XP_005248074.1
10
1106
XM_005248018.4
XP_005248075.1
10
1105
XM_005248019.4
XP_005248076.1
10
1105
XM_005248020.4
XP_005248077.1
10
1105
XM_005248021.4
XP_005248078.1
10
1104
XM_005248022.4
XP_005248079.1
10
1105
XM_005248023.4
XP_005248080.1
10
1105
XM_005248024.4
XP_005248081.1
10
1104
XM_005248027.4
XP_005248084.1
10
1104
XM_005248028.4
XP_005248085.1
10
1101
XM_005248029.4
XP_005248086.1
10
1103
XM_005248030.4
XP_005248087.1
10
1103
XM_005248031.4
XP_005248088.1
10
1105
XM_006713923.3
XP_006713986.1
10
1106
XM_006713924.3
XP_006713987.1
10
1105
XM_006713925.3
XP_006713988.1
10
1106
XM_006713927.3
XP_006713990.1
10
1104
XM_011513586.2
XP_011511888.1
10
1110
XM_017008706.1
XP_016864195.1
10
1107
XM_017008707.1
XP_016864196.1
10
1116
XM_017008708.1
XP_016864197.1
10
1105
XM_017008709.1
XP_016864198.1
10
1106
XM_017008710.1
XP_016864199.1
10
1105
XM_017008711.1
XP_016864200.1
10
1105
XM_017008712.1
XP_016864201.1
10
1105
XM_017008713.1
XP_016864202.1
10
1104
XM_017008714.1
XP_016864203.1
10
1116
XM_017008715.1
XP_016864204.1
10
1105
XM_017008716.1
XP_016864205.1
10
1104
XM_017008717.1
XP_016864206.1
10
1104
XM_017008718.1
XP_016864207.1
10
1116
XM_017008719.1
XP_016864208.1
10
1104
XM_017008720.1
XP_016864209.1
10
1103
XM_017008721.1
XP_016864210.1
10
1103
XM_017008722.1
XP_016864211.1
10
1104
XM_017008723.1
XP_016864212.1
10
1116
XM_017008724.1
XP_016864213.1
10
1116
XM_017008725.1
XP_016864214.1
10
1116
XM_017008726.1
XP_016864215.1
5
565
XM_017008727.1
XP_016864216.1
5
565
GenBank mRNA
AB058711.1
-
9
894
AJ748600.1
-
10
1057
AJ748601.1
-
10
1057
AK094754.1
-
10
1114
AK094798.1
-
5
535
AK301060.1
-
10
1098
BC067214.1
-
10
1147
BC122567.1
-
10
1086
40085570
BE155865.1
-
2
173
DQ413176.1
-
10
1012
DQ413177.1
-
9
992
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
actin cytoskeleton organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline