Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271286.1 | NP_001258215.1 | 2 | 223 | ||
NM_001271289.1 | NP_001258218.1 | 2 | 209 | |||
NM_001271318.1 | NP_001258247.1 | 3 | 324 | |||
NM_032434.3 | NP_115810.2 | 2 | 223 | |||
GenBank mRNA | AK293771.1 | - | 2 | 123 | ||
AK294055.1 | - | 2 | 332 | |||
AK299818.1 | - | 2 | 113 | |||
AK316011.1 | - | 3 | 184 | |||
AL514772.3 | - | 2 | 140 | |||
BG720805.1 | - | 2 | 173 | |||
BP251717.1 | - | 2 | 223 |