Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271286.1 | NP_001258215.1 | 6 | 696 | ||
NM_001271287.1 | NP_001258216.1 | 6 | 724 | |||
NM_001271288.1 | NP_001258217.1 | 6 | 721 | |||
NM_001271289.1 | NP_001258218.1 | 6 | 685 | |||
NM_001271318.1 | NP_001258247.1 | 8 | 884 | |||
NM_032434.3 | NP_115810.2 | 7 | 783 | |||
GenBank mRNA | AB058708.1 | - | 5 | 542 | ||
AK057028.1 | - | 7 | 693 | |||
AK293771.1 | - | 6 | 599 | |||
AK294055.1 | - | 7 | 889 | |||
AK299818.1 | - | 6 | 586 | |||
AK316011.1 | - | 8 | 744 | |||
AK316299.1 | - | 6 | 638 | |||
AL514772.3 | - | 7 | 700 | |||
BC043221.2 | - | 6 | 632 | 5295402 | ||
BC093041.1 | - | 6 | 651 | 6173398 | ||
BX647996.1 | - | 6 | 630 |