Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011657.3 | NP_001011657.2 | 7 | 1191 | ||
NM_001282400.1 | NP_001269329.1 | 10 | 1747 | |||
NM_001282401.1 | NP_001269330.1 | 9 | 1694 | |||
XM_005262212.4 | XP_005262269.2 | 7 | 1208 | |||
XM_005262213.4 | XP_005262270.2 | 7 | 1205 | |||
XM_005262214.3 | XP_005262271.2 | 6 | 1139 | |||
XM_005262216.4 | XP_005262273.2 | 7 | 910 | |||
XM_006724711.3 | XP_006724774.1 | 3 | 670 | |||
XM_017029903.1 | XP_016885392.1 | 7 | 907 | |||
XM_017029904.1 | XP_016885393.1 | 7 | 2960 | |||
GenBank mRNA | AB058692.1 | - | 2 | 3824 | ||
AK094141.1 | - | 10 | 1708 | |||
AK296520.1 | - | 9 | 1659 | |||
BC140920.1 | - | 2 | 3790 | 9056732 | ||
BC152470.1 | - | 2 | 3824 | 8862607 |