Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146028.1 | NP_001139500.1 | 6 | 2997 | ||
NM_032452.2 | NP_115828.2 | 7 | 3123 | |||
GenBank mRNA | AB058734.1 | - | 6 | 2997 | ||
AK055345.1 | - | 3 | 1021 | |||
AL834411.1 | - | 4 | 1207 | |||
AY344068.1 | - | 5 | 2220 | |||
BC055429.1 | - | 7 | 3108 | 5262644 |