Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145459.1 | NP_001138931.1 | 3 | 909 | ||
NM_001145460.1 | NP_001138932.1 | 5 | 1393 | |||
NM_032495.5 | NP_115884.4 | 4 | 1263 | |||
NM_139211.4 | NP_631957.1 | 3 | 865 | |||
NM_139212.3 | NP_631958.1 | 3 | 1138 | |||
XM_017008728.1 | XP_016864217.1 | 3 | 900 | |||
XM_017008729.1 | XP_016864218.1 | 3 | 782 | |||
XM_017008730.1 | XP_016864219.1 | 3 | 830 | |||
XM_017008731.1 | XP_016864220.1 | 3 | 865 | |||
XM_017008732.1 | XP_016864221.1 | 4 | 1141 | |||
XM_017008733.1 | XP_016864222.1 | 3 | 705 | |||
XM_017008734.1 | XP_016864223.1 | 3 | 761 | |||
GenBank mRNA | AB059408.1 | - | 3 | 823 | ||
AB059409.1 | - | 3 | 706 | |||
AB059410.2 | - | 3 | 715 | |||
AF454763.1 | - | 3 | 729 | |||
AF492675.1 | - | 3 | 1113 | |||
AF492676.1 | - | 4 | 845 | |||
AF492677.1 | - | 4 | 889 | |||
AF492678.1 | - | 5 | 1019 | |||
AF492679.1 | - | 5 | 975 | |||
AF492680.1 | - | 3 | 865 | |||
AF492681.1 | - | 3 | 828 | |||
AK123318.1 | - | 1 | 2935 | |||
BC014225.2 | - | 2 | 978 | 4335211 |