Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129993.1 | NP_001123465.1 | 13 | 1752 | ||
NM_032506.2 | NP_115895.2 | 13 | 1826 | |||
XM_005264605.3 | XP_005264662.1 | 13 | 1753 | |||
XM_005264606.4 | XP_005264663.1 | 13 | 1753 | |||
XM_005264607.2 | XP_005264664.1 | 12 | 1639 | |||
XM_011533126.2 | XP_011531428.1 | 13 | 1805 | |||
XM_011533127.2 | XP_011531429.1 | 13 | 1665 | |||
XM_011533128.1 | XP_011531430.1 | 13 | 1753 | |||
XM_011533129.2 | XP_011531431.1 | 12 | 1600 | |||
XM_011533131.2 | XP_011531433.1 | 13 | 1753 | |||
XM_017005108.1 | XP_016860597.1 | 13 | 1667 | |||
XM_017005109.1 | XP_016860598.1 | 13 | 1753 | |||
GenBank mRNA | AB058744.1 | - | 8 | 864 | ||
AK309749.1 | - | 13 | 1734 | |||
AL833595.1 | - | 13 | 1692 | |||
BC039298.1 | - | 12 | 1649 | 5314819 | ||
BC070104.1 | - | 13 | 1826 | 5268265 | ||
BX647395.1 | - | 12 | 1449 |