Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308396.1 | NP_001295325.1 | 3 | 485 | ||
NM_032112.2 | NP_115488.2 | 3 | 481 | |||
NM_176792.2 | NP_789762.1 | 3 | 481 | |||
NM_176793.1 | NP_789763.1 | 3 | 481 | |||
NM_176794.1 | NP_789764.1 | 3 | 481 | |||
XM_005270231.2 | XP_005270288.1 | 3 | 476 | |||
XM_006718035.3 | XP_006718098.1 | 3 | 476 | |||
XM_017016790.1 | XP_016872279.1 | 2 | 593 | |||
GenBank mRNA | AB049656.1 | - | 3 | 443 | ||
AK095556.1 | - | 3 | 465 | |||
AK291069.1 | - | 3 | 436 | |||
AK300237.1 | - | 3 | 463 | |||
BC015905.1 | - | 3 | 481 | 3921157 | ||
BC031287.1 | - | 2 | 368 | |||
BC041165.1 | - | 3 | 415 | 5590251 | ||
BC052639.1 | - | 3 | 422 | 6501303 | ||
BC053373.1 | - | 3 | 434 | 6020112 | ||
BU597385.1 | - | 3 | 486 |