Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM185A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174092.2 | NP_001167563.1 | 6 | 1119 | ||
NM_032508.3 | NP_115897.1 | 7 | 1296 | |||
NR_104121.1 | 4 | 827 | ||||
GenBank mRNA | AF353675.1 | - | 4 | 523 | ||
AF530473.1 | - | 8 | 1275 | |||
AK096308.1 | - | 7 | 1108 | |||
AK097391.1 | - | 4 | 633 | |||
AK128688.1 | - | 3 | 2198 | |||
AK297335.1 | - | 6 | 982 | |||
BC013793.1 | - | 3 | 448 | |||
BC103821.1 | - | 7 | 1000 | 40000939 | ||
BC103822.1 | - | 7 | 1000 | 40000940 |