Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080744.1 | NP_001074213.1 | 4, 5 | 853 | ||
NM_001080745.1 | NP_001074214.1 | 4, 5 | 853 | |||
NM_001346.2 | NP_001337.2 | 4, 5 | 853 | |||
GenBank mRNA | AB209033.1 | - | 4, 5 | 696 | ||
AK304440.1 | - | 4, 5 | 851 | |||
AK314192.1 | - | 4, 5 | 798 | |||
AY737485.1 | - | 3, 4 | 548 | |||
BC089411.1 | - | 4, 5 | 607 | 3081161 | ||
BC112363.1 | - | 3, 4 | 576 | 40054027 | ||
D26135.1 | - | 4, 5 | 832 |