Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142351.1 | NP_001135823.1 | 5 | 1315 | ||
NM_001142352.1 | NP_001135824.1 | 5 | 1385 | |||
NM_001322362.1 | NP_001309291.1 | 5 | 1364 | |||
NM_032528.2 | NP_115917.1 | 5 | 1385 | |||
NR_136313.1 | 4 | 400 | ||||
XM_006712802.1 | XP_006712865.1 | 5 | 1400 | |||
XM_011511999.2 | XP_011510301.1 | 5 | 1497 | |||
XM_011512000.2 | XP_011510302.1 | 5 | 1582 | |||
XM_011512001.2 | XP_011510303.1 | 5 | 1582 | |||
XM_017005110.1 | XP_016860599.1 | 5 | 1336 | |||
GenBank mRNA | AB058780.1 | - | 5 | 1315 | ||
AB059555.1 | - | 5 | 1371 | |||
AJ512141.1 | - | 3 | 256 | |||
AK307784.1 | - | 5 | 1385 | |||
BC008680.1 | - | 5 | 1371 | 3856171 |