Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080744.1 | NP_001074213.1 | 4, 5 | 848 | ||
NM_001080745.1 | NP_001074214.1 | 4, 5 | 848 | |||
NM_001346.2 | NP_001337.2 | 4, 5 | 848 | |||
GenBank mRNA | AB209033.1 | - | 4, 5 | 691 | ||
AK304440.1 | - | 4, 5 | 846 | |||
AK314192.1 | - | 4, 5 | 793 | |||
AY737485.1 | - | 3, 4 | 543 | |||
BC089411.1 | - | 4, 5 | 602 | 3081161 | ||
BC112363.1 | - | 3, 4 | 571 | 40054027 | ||
D26135.1 | - | 4, 5 | 827 |