Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161707.1 | NP_001155179.1 | 8 | 1278 | ||
NM_001301097.1 | NP_001288026.1 | 8 | 1278 | |||
NM_032531.3 | NP_115920.1 | 8 | 1278 | |||
XM_011543026.2 | XP_011541328.1 | 8 | 1022 | |||
XM_011543027.2 | XP_011541329.1 | 9 | 1483 | |||
XM_011543028.2 | XP_011541330.1 | 9 | 1342 | |||
XM_011543030.2 | XP_011541332.1 | 8 | 1019 | |||
XM_011543031.2 | XP_011541333.1 | 9 | 1477 | |||
XM_011543032.2 | XP_011541334.1 | 8 | 1019 | |||
XM_011543033.1 | XP_011541335.1 | 9 | 1483 | |||
XM_017018418.1 | XP_016873907.1 | 8 | 1573 | |||
XM_017018419.1 | XP_016873908.1 | 9 | 1483 | |||
XM_017018420.1 | XP_016873909.1 | 9 | 1342 | |||
GenBank mRNA | AB058770.1 | - | 8 | 1167 | ||
AF480410.1 | - | 8 | 917 | |||
AY358743.1 | - | 8 | 1218 | |||
BC101775.1 | - | 8 | 1003 | 8069281 | ||
BC101801.1 | - | 8 | 1003 | 8069307 |