Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032532.2 | NP_115921.2 | 4 | 609 | ||
GenBank mRNA | AB058769.2 | - | 3 | 284 | ||
BC146783.1 | - | 3 | 284 | 9007239 | ||
BC150607.1 | - | 3 | 284 | 9056977 | ||
DQ009660.1 | - | 4 | 609 |