Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290187.1 | NP_001277116.1 | 5 | 559 | ||
NM_032534.3 | NP_115923.2 | 5 | 858 | |||
XM_011516636.2 | XP_011514938.1 | 6 | 738 | |||
XM_011516637.2 | XP_011514939.1 | 6 | 735 | |||
XM_011516638.2 | XP_011514940.1 | 6 | 694 | |||
XM_011516639.2 | XP_011514941.1 | 6 | 738 | |||
XM_011516640.2 | XP_011514942.1 | 6 | 735 | |||
XM_011516641.2 | XP_011514943.1 | 5 | 626 | |||
XM_011516642.2 | XP_011514944.1 | 5 | 778 | |||
XM_011516643.2 | XP_011514945.1 | 5 | 639 | |||
GenBank mRNA | AB058765.2 | - | 5 | 812 | ||
AL831935.1 | - | 3 | 283 | |||
BC142722.1 | - | 5 | 559 | 40148879 | ||
BC146667.1 | - | 5 | 559 | 40148146 | ||
BC150330.1 | - | 5 | 823 | 8860497 | ||
BC152409.1 | - | 5 | 812 | 8862512 |