Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032536.2 | NP_115925.2 | 3 | 1062 | ||
XM_006717304.3 | XP_006717367.1 | 5 | 1083 | |||
XM_011519094.2 | XP_011517396.1 | 5 | 1083 | |||
XM_011519096.2 | XP_011517398.1 | 5 | 1480 | |||
XM_011519097.2 | XP_011517399.1 | 4 | 1001 | |||
XM_011519098.2 | XP_011517400.1 | 5 | 1033 | |||
XM_011519099.2 | XP_011517401.1 | 4 | 1004 | |||
XM_011519100.2 | XP_011517402.1 | 3 | 933 | |||
XM_011519102.2 | XP_011517404.1 | 4 | 948 | |||
XM_011519103.2 | XP_011517405.1 | 3 | 840 | |||
XM_011519104.2 | XP_011517406.1 | 3 | 1064 | |||
XM_011519105.2 | XP_011517407.1 | 3 | 930 | |||
XM_011519106.2 | XP_011517408.1 | 5 | 1083 | |||
XM_011519107.2 | XP_011517409.1 | 5 | 1083 | |||
XM_011519108.2 | XP_011517410.1 | 5 | 1083 | |||
XM_011519109.2 | XP_011517411.1 | 5 | 1083 | |||
XM_011519110.2 | XP_011517412.1 | 5 | 1083 | |||
XM_017015212.1 | XP_016870701.1 | 6 | 1187 | |||
XM_017015213.1 | XP_016870702.1 | 4 | 1390 | |||
XM_017015214.1 | XP_016870703.1 | 5 | 1083 | |||
XM_017015216.1 | XP_016870705.1 | 5 | 1083 | |||
GenBank mRNA | AB058760.2 | - | 3 | 1062 | ||
AY358165.1 | - | 4 | 632 | |||
BC013770.1 | - | 2 | 371 | 4344617 |